Assay Details
| Gene Symbol: TEFM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001168010.2 | NP_001161482.2 | 2 - 3 | 537 | 107 | |
| GenBank mRNA | BC149452.1 | - | 2 - 3 | 522 | 107 | |
More Information
Set Membership: |
|

TEFM
Genomic Map