Assay Details
| Gene Symbol: TEFM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001168010.2 | NP_001161482.2 | 3 - 4 | 685 | 96 | |
| GenBank mRNA | BC149452.1 | - | 3 - 4 | 670 | 96 | |
More Information
Set Membership: |
|

TEFM
Genomic Map