Assay Details
| Gene Symbol: LIG4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191126.1 | NP_001178055.1 | 1 - 2 | 73 | 84 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001191126.1 transcripts.
Set Membership: |
|

LIG4
Genomic Map