Assay Details
| Gene Symbol: RNF43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191194.1 | NP_001178123.1 | 6 - 7 | 846 | 75 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

RNF43| Gene Symbol: RNF43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191194.1 | NP_001178123.1 | 6 - 7 | 846 | 75 | |
Set Membership: |
|