Assay Details
Gene Symbol: RBAK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191256.1 | NP_001178185.1 | 1 - 2 | 492 | 103 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001191256.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
