Assay Details
| Gene Symbol: SPACA7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001207045.1 | NP_001193974.1 | 1 - 2 | 136 | 78 | |
| GenBank mRNA | DT848094.1 | - | 1 - 2 | 106 | 78 | |
More Information
Set Membership: |
|

SPACA7
Genomic Map