Assay Details
| Gene Symbol: SPACA7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001207045.1 | NP_001193974.1 | 3 - 4 | 296 | 76 | |
| GenBank mRNA | BC108171.1 | - | 3 - 4 | 297 | 76 | |
| DT848094.1 | - | 3 - 4 | 266 | 76 | ||
More Information
Set Membership: |
|

SPACA7
Genomic Map