Assay Details
| Gene Symbol: ANKRD34B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001207060.1 | NP_001193989.1 | 3 - 4 | 305 | 102 | |
| GenBank mRNA | BC122789.1 | - | 3 - 4 | 306 | 102 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC122789.1, NM_001207060.1 transcripts.
Set Membership: |
|

ANKRD34B
Genomic Map