Assay Details
| Gene Symbol: CRNN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191303.1 | NP_001178232.1 | 2 - 3 | 767 | 98 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CRNN| Gene Symbol: CRNN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191303.1 | NP_001178232.1 | 2 - 3 | 767 | 98 | |
Set Membership: |
|