Assay Details
| Gene Symbol: SERPINA7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174668.2 | NP_777093.1 | 2 - 3 | 659 | 74 | |
| GenBank mRNA | AF204928.1 | - | 2 - 3 | 659 | 74 | |
| BC103463.1 | - | 2 - 3 | 660 | 74 | ||
More Information
Set Membership: |
|

SERPINA7
Genomic Map