Assay Details
Gene Symbol: ST6GALNAC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205791.1 | NP_991360.1 | 3 - 4 | 713 | 83 | |
GenBank mRNA | AJ620651.1 | - | 3 - 4 | 713 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
