Assay Details
Gene Symbol: GCM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192105.1 | NP_001179034.1 | 3 - 4 | 528 | 52 | |
NM_001205290.1 | NP_001192219.1 | 4 - 5 | 622 | 52 | ||
GenBank mRNA | AY173016.1 | - | 1 - 2 | 185 | 52 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
