Assay Details
| Gene Symbol: FGF20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001192165.1 | NP_001179094.1 | 2 - 3 | 388 | 88 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

FGF20| Gene Symbol: FGF20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001192165.1 | NP_001179094.1 | 2 - 3 | 388 | 88 | |
Set Membership: |
|