Assay Details
| Gene Symbol: DSG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001192172.2 | NP_001179101.2 | 1 - 2 | 36 | 65 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

DSG2| Gene Symbol: DSG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001192172.2 | NP_001179101.2 | 1 - 2 | 36 | 65 | |
Set Membership: |
|