Assay Details
Gene Symbol: LY6G5B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192202.1 | NP_001179131.1 | 2 - 3 | 179 | 75 | |
GenBank mRNA | HE864490.1 | - | 1 - 2 | 330 | 75 |
Gene Symbol: CSNK2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | HE864424.1 | - | 7 - 8 | 803 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
