Assay Details
| Gene Symbol: ZNF432 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001192232.2 | NP_001179161.2 | 1 - 2 | 125 | 60 | |
| GenBank mRNA | BC151729.1 | - | 1 - 2 | 126 | 60 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC151729.1, NM_001192232.2 transcripts.
Set Membership: |
|

ZNF432
Genomic Map