Assay Details
| Gene Symbol: ZNF432 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001192232.2 | NP_001179161.2 | 4 - 5 | 481 | 102 | |
| GenBank mRNA | BC151729.1 | - | 4 - 5 | 482 | 102 | |
More Information
Set Membership: |
|

ZNF432
Genomic Map