Assay Details
| Gene Symbol: PITX3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001192305.1 | NP_001179234.1 | 1 - 2 | 181 | 77 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001192305.1 transcripts.
Set Membership: |
|

PITX3
Genomic Map