Assay Details
| Gene Symbol: CTDSP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001192440.1 | NP_001179369.1 | 1 - 2 | 182 | 115 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CTDSP1| Gene Symbol: CTDSP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001192440.1 | NP_001179369.1 | 1 - 2 | 182 | 115 | |
Set Membership: |
|