Assay Details
| Gene Symbol: TMEM150A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193030.1 | NP_001179959.1 | 4 - 5 | 379 | 92 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TMEM150A| Gene Symbol: TMEM150A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193030.1 | NP_001179959.1 | 4 - 5 | 379 | 92 | |
Set Membership: |
|