Assay Details
Gene Symbol: SULT1C3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193080.1 | NP_001180009.1 | 2 - 3 | 303 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
