Assay Details
| Gene Symbol: CEPT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193130.1 | NP_001180059.1 | 2 - 3 | 492 | 69 | |
| GenBank mRNA | BC112673.1 | - | 2 - 3 | 474 | 69 | |
More Information
Set Membership: |
|

CEPT1
Genomic Map