Assay Details
| Gene Symbol: FAM229B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195067.1 | NP_001181996.1 | 3 - 4 | 451 | 66 | |
| GenBank mRNA | BC123382.1 | - | 3 - 4 | 453 | 66 | |
More Information
Set Membership: |
|

FAM229B
Genomic Map