Assay Details
| Gene Symbol: VPS37B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205357.1 | NP_001192286.1 | 1 - 2 | 131 | 67 | |
| GenBank mRNA | BC149169.1 | - | 1 - 2 | 104 | 67 | |
| EE392225.1 | - | 1 - 2 | 131 | 67 | ||
More Information
Set Membership: |
|

VPS37B
Genomic Map