Assay Details
| Gene Symbol: VPS37B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205357.1 | NP_001192286.1 | 3 - 4 | 380 | 117 | |
| GenBank mRNA | BC149169.1 | - | 3 - 4 | 353 | 117 | |
| EE392225.1 | - | 3 - 4 | 340 | 117 | ||
More Information
Set Membership: |
|

VPS37B
Genomic Map