Assay Details
| Gene Symbol: PLD5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205378.1 | NP_001192307.1 | 5 - 6 | 735 | 64 | |
| GenBank mRNA | BC150069.1 | - | 5 - 6 | 713 | 64 | |
More Information
Set Membership: |
|

PLD5
Genomic Map