Assay Details
| Gene Symbol: RASD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001206261.2 | NP_001193190.1 | 1 - 2 | 417 | 107 | |
| GenBank mRNA | BC103301.1 | - | 1 - 2 | 285 | 107 | |
More Information
Set Membership: |
|

RASD1
Genomic Map