Assay Details
| Gene Symbol: CYP8B1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076139.2 | NP_001069607.1 | 2 | 922 | 128 | |
| GenBank mRNA | BC112645.1 | - | 2 | 922 | 128 | |
More Information
Set Membership: |
|

CYP8B1
Genomic Map