Assay Details
| Gene Symbol: HSPA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174344.1 | NP_776769.1 | 1 | 1819 | 107 | |
| GenBank mRNA | BC105156.1 | - | 1 | 1949 | 107 | |
| BT030673.1 | - | 1 | 1923 | 107 | ||
More Information
Set Membership: |
|

HSPA2
Genomic Map