Assay Details
Gene Symbol: MIR2917 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_031294.1 | 1 | 46 | 72 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | collagen type V alpha 2 chain | COL5A2 | Sense | Chr. 2 - 7198093 - 7358813 on NCBI Build ARS-UCD1.2 | ||
Downstream | collagen type III alpha 1 chain | COL3A1 | 102.3kb | Anti-sense | Chr. 2 - 7375278 - 7414912 on NCBI Build ARS-UCD1.2 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
