Assay Details
| Gene Symbol: MIR2402 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_031306.1 | 1 | 36 | 68 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | cytochrome c oxidase assembly factor 6 homolog (S. cerevisiae) | COA6 | C28H1orf31 | -638.8kb | Sense | Chr. 28 - 7188602 - 7197787 on NCBI Build ARS-UCD1.2 |
| Downstream | translocase of outer mitochondrial membrane 20 | TOMM20 | 46.7kb | Anti-sense | Chr. 28 - 7883316 - 7898012 on NCBI Build ARS-UCD1.2 |
More Information
Set Membership: |
|

MIR2402
Genomic Map