Assay Details
| Gene Symbol: MIR210 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_049717.1 | 1 | 49 | 58 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | PHD and ring finger domains 1 | PHRF1 | -7.1kb | Anti-sense | Chr. 29 - 50699012 - 50721935 on NCBI Build ARS-UCD1.2 | |
| Downstream | Ras association domain family member 7 | RASSF7 | 2.9kb | Anti-sense | Chr. 29 - 50732080 - 50734636 on NCBI Build ARS-UCD1.2 |
More Information
Set Membership: |
|

MIR210
Genomic Map