Assay Details
| Gene Symbol: MIR1282 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_030822.1 | 1 | 43 | 84 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | small EDRK-rich factor 2 | SERF2 | Anti-sense | Chr. 21 - 55412552 - 55416413 on NCBI Build ARS-UCD1.2 | ||
| Downstream | huntingtin interacting protein K | HYPK | C21H15orf63 | 6.5kb | Anti-sense | Chr. 21 - 55420598 - 55421853 on NCBI Build ARS-UCD1.2 |
More Information
Set Membership: |
|

MIR1282
Genomic Map