Assay Details
| Gene Symbol: MIR124A-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_031373.1 | 1 | 38 | 84 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | methionine sulfoxide reductase A | MSRA | -132.0kb | Anti-sense | Chr. 8 - 8786080 - 9169884 on NCBI Build ARS-UCD1.2 | |
| Downstream | homeobox containing 1 | HMBOX1 | 347.6kb | Anti-sense | Chr. 8 - 9649607 - 9840092 on NCBI Build ARS-UCD1.2 |
More Information
Set Membership: |
|

MIR124A-1
Genomic Map