Assay Details
| Gene Symbol: ASB12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001192225.3 | NP_001179154.1 | 1 - 2 | 853 | 99 | |
| GenBank mRNA | BC122821.1 | - | 1 - 2 | 1292 | 99 | |
More Information
Set Membership: |
|

ASB12
Genomic Map