Assay Details
| Gene Symbol: TMEM95 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001367291.1 | NP_001354220.1 | 7 | 65 | ||
| GenBank mRNA | KT200428.1 | - | 6 | 526 | 65 | |
| KT200429.1 | - | 3 | 296 | 65 | ||
More Information
Set Membership: |
|

TMEM95
Genomic Map