Assay Details
| Gene Symbol: acd-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001270321.3 | NP_001257250.1 | 11 - 12 | 1336 | 87 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

acd-3| Gene Symbol: acd-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001270321.3 | NP_001257250.1 | 11 - 12 | 1336 | 87 | |
Set Membership: |
|