Assay Details
| Gene Symbol: mfsd-10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_078413.5 | NP_510814.1 | 12 - 13 | 1319 | 92 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

mfsd-10| Gene Symbol: mfsd-10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_078413.5 | NP_510814.1 | 12 - 13 | 1319 | 92 | |
Set Membership: |
|