Assay Details
| Gene Symbol: F46F11.1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_170868.5 | NP_740855.2 | 11 - 12 | 2233 | 153 | |
| NM_171837.9 | NP_740856.1 | 11 - 12 | 2239 | 153 | ||
More Information
Set Membership: |
|

F46F11.1
Genomic Map