Assay Details
| Gene Symbol: nhx-8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026557.4 | NP_001021728.1 | 3 - 4 | 365 | 63 | |
| NM_001026558.5 | NP_001021729.1 | 2 - 3 | 234 | 63 | ||
| GenBank mRNA | AF497832.1 | - | 3 - 4 | 314 | 63 | |
| AF497833.1 | - | 2 - 3 | 256 | 63 | ||
More Information
Set Membership: |
|

nhx-8
Genomic Map