Assay Details
Gene Symbol: spe-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_059694.6 | NP_492095.1 | 2 - 3 | 513 | 110 | |
GenBank mRNA | Z14066.1 | - | 474 | 110 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
