Assay Details
| Gene Symbol: mgl-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001313467.3 | NP_001300396.1 | 3 | 443 | 79 | |
| NM_001313468.3 | NP_001300397.1 | 3 - 4 | 522 | 79 | ||
| NM_060319.5 | NP_492720.3 | 3 - 4 | 515 | 79 | ||
| Gene Symbol: maf-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001381197.2 | NP_001367180.1 | 3 - 4 | 520 | 79 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001381197.2 transcripts.
Set Membership: |
|

mgl-2
Genomic Map