Assay Details
Gene Symbol: spe-15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001306501.3 | NP_001293430.1 | 7 - 8 | 1133 | 79 | |
NM_001306502.3 | NP_001293431.1 | 3 - 4 | 599 | 79 | ||
NM_058455.5 | NP_490856.1 | 14 - 15 | 3249 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
