Assay Details
Gene Symbol: spe-15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001306501.3 | NP_001293430.1 | 9 - 10 | 1384 | 92 | |
NM_001306502.3 | NP_001293431.1 | 5 - 6 | 850 | 92 | ||
NM_058455.5 | NP_490856.1 | 16 - 17 | 3500 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
