Assay Details
| Gene Symbol: F46F11.1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_171837.9 | NP_740856.1 | 17 - 18 | 3195 | 72 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

F46F11.1| Gene Symbol: F46F11.1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_171837.9 | NP_740856.1 | 17 - 18 | 3195 | 72 | |
Set Membership: |
|