Assay Details
| Gene Symbol: ces-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_061209.6 | NP_493610.1 | 1 - 2 | 309 | 82 | |
| Gene Symbol: ZK909.6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026711.4 | NP_001021882.2 | 1 - 2 | 305 | 82 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001026711.4 transcripts.
Set Membership: |
|

ces-2
Genomic Map