Assay Details
| Gene Symbol: abhd-5.2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_071896.6 | NP_504297.1 | 2 - 3 | 403 | 104 | |
| NM_182378.4 | NP_872178.1 | 1 - 2 | 155 | 104 | ||
More Information
Set Membership: |
|

abhd-5.2
Genomic Map