Assay Details
Gene Symbol: sra-26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_062951.3 | NP_495352.1 | 3 - 4 | 670 | 150 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
