Assay Details
| Gene Symbol: jun-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027195.4 | NP_001022366.1 | 3 - 4 | 441 | 91 | |
| NM_001027196.5 | NP_001022367.1 | 3 - 4 | 306 | 91 | ||
| NM_001356878.2 | NP_001343785.1 | 2 - 3 | 304 | 91 | ||
| NM_001356879.3 | NP_001343787.1 | 1 - 2 | 109 | 91 | ||
| NM_001383910.1 | 3 - 4 | 364 | 91 | |||
| GenBank mRNA | EU553920.1 | - | 3 - 4 | 379 | 91 | |
| EU553922.1 | - | 1 - 2 | 145 | 91 | ||
| Gene Symbol: C16D2.1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001381554.1 | NP_001367159.1 | 3 - 4 | 464 | 91 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001381554.1, NM_001383910.1 transcripts.
Set Membership: |
|

jun-1
Genomic Map