Assay Details
| Gene Symbol: jun-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027197.5 | NP_001022368.1 | 1 - 2 | 134 | 78 | |
| GenBank mRNA | EU553925.1 | - | 1 - 2 | 107 | 78 | |
More Information
Set Membership: |
|

jun-1
Genomic Map